Please login to your online account to display your discounted pricing

Chlorobenzene ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Chlorobenzene
CAS 108-90-7
Purity Grade ACS Reagent Grade
Molecular Formula C6H5Cl
Formula Weight 112.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK44194

 
avantor performance
441908
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK4419500

 
avantor performance
441904
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 112.56
Quantity 4L
Packaging Glass
Molecular Formula C6H5Cl
CAS 108-90-7
Chemical Name or Material Chlorobenzene