Learn More
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036401M1
Description
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chlorobenzene | |
108-90-7 | |
1.106 g/mL | |
4 x 1 L | |
Aromatic | |
1.524 | |
MFCD00000530 | |
605632 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
chlorobenzene | |
7964 | |
112.56 | |
ACS |
-45°C | |
Colorless | |
132°C | |
28°C (82°F) | |
Liquid | |
C6H5Cl | |
UN1134 | |
14,2121 | |
Miscible with methanol and diethyl ether. Immiscible with water. | |
C1=CC=C(C=C1)Cl | |
112.556 | |
CHEBI:28097 | |
99.5% |
Chemical Identifiers
108-90-7 | |
112.556 | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
7964 | |
chlorobenzene |
C6H5Cl | |
MFCD00000530 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
CHEBI:28097 | |
C1=CC=C(C=C1)Cl |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
EINECSNumber : 203-628-5
RTECSNumber : CZ0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only