Please login to your online account to display your discounted pricing

Chlorobenzene, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC396971000

 View more versions of this product

Catalog No. AC396971000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Physical Form Liquid
Formula Weight 112.56
Quantity 100mL
Color Clear colorless
Merck Index 15, 2122
Packaging AcroSeal™ Glass Bottle
Density 1.1075g/mL
Purity Grade Extra Dry
Water 0.005% Max. (Coulometric)
Additional Information Filtered over 0.2 micrometer
Chemical Name or Material Chlorobenzene
Name Note Extra Dry
Assay Percent Range 99.8%
CAS 108-90-7
Molecular Formula C6H5Cl
Health Hazard 1 Warning
MDL Number MFCD00000530
UN Number 1134
Residue after Evaporation 0.001% max.
Linear Formula C6H5Cl
Health Hazard 2 GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Health Hazard 3 GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Specifications

Chemical Name or Material Chlorobenzene
Name Note Extra Dry
CAS 108-90-7
Purity Grade Extra Dry
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula C6H5Cl
Linear Formula C6H5Cl
Formula Weight 112.56
MDL Number MFCD00000530
Accessories

Warning

GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.

SDS & Safety Documents