Please login to your online account to display your discounted pricing

Chloroacetyl isocyanate, 90%, Tech., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314300010

 View more versions of this product

Catalog No. AC314300010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥88%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 119.51
Quantity 1g
Color Clear yellow
EINECS Number 224-715-4
Density 1.403g/mL
Merck Index 15, 2117
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Chloroacetyl isocyanate
Name Note tech., 90%
Assay Percent Range 90%
Molecular Formula C3H2ClNO2
Linear Formula ClCHCONCO
CAS 4461-30-7
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00002034
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

Specifications

Chemical Name or Material Chloroacetyl isocyanate
Name Note tech., 90%
CAS 4461-30-7
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C3H2ClNO2
Linear Formula ClCHCONCO
Formula Weight 119.51
MDL Number MFCD00002034
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

SDS & Safety Documents