missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetyl isocyanate, 90%, Tech., ACROS Organics™

$44.72 - $191.77

Chemical Identifiers

CAS 4461-30-7
Molecular Formula C3H2ClNO2
Molecular Weight (g/mol) 119.51
MDL Number MFCD00002034
InChI Key MOVMEFHWBOWMFU-UHFFFAOYSA-N
Synonym acetyl isocyanate, chloro, acmc-20alh0, alpha-chloroacetyl isocyanate, chloro-acetyl isocyanate, chloroacetyl isocyanate, chloroacetyl isocyante, chloroacetylisocyanate, h5wf2amh7j, snxhcfhai zyp@, unii-h5wf2amh7j
PubChem CID 78209
IUPAC Name 2-chloroacetyl isocyanate
SMILES C(C(=O)N=C=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC314300010
SDS View Documents
Thermo Fisher Scientific
AC314300010
1g Glass bottle
Each for $44.72
 
AC314300050
SDS View Documents
Thermo Fisher Scientific
314300050
5g Glass bottle
Each for $191.77
 
Description

Description

Specifications

Chemical Identifiers

4461-30-7
119.51
MOVMEFHWBOWMFU-UHFFFAOYSA-N
78209
C(C(=O)N=C=O)Cl
C3H2ClNO2
MFCD00002034
acetyl isocyanate, chloro, acmc-20alh0, alpha-chloroacetyl isocyanate, chloro-acetyl isocyanate, chloroacetyl isocyanate, chloroacetyl isocyante, chloroacetylisocyanate, h5wf2amh7j, snxhcfhai zyp@, unii-h5wf2amh7j
2-chloroacetyl isocyanate

Specifications

1.4030g/mL
Authentic
1.403
Yellow
4461-30-7
C3H2ClNO2
MFCD00002034
acetyl isocyanate, chloro, acmc-20alh0, alpha-chloroacetyl isocyanate, chloro-acetyl isocyanate, chloroacetyl isocyanate, chloroacetyl isocyante, chloroacetylisocyanate, h5wf2amh7j, snxhcfhai zyp@, unii-h5wf2amh7j
C(C(=O)N=C=O)Cl
119.51
119.51
90%
Chloroacetyl isocyanate, tech., 90%
61°C
Glass bottle
50.0°C to 55.0°C (20.0mmHg)
1g
90%
ClCH2CONCO
15, 2117
MOVMEFHWBOWMFU-UHFFFAOYSA-N
2-chloroacetyl isocyanate
78209
Liquid
Technical
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-715-4

SDS