missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetonitrile, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC108521000

 View more versions of this product

Catalog No. AC108521000


 

Specifications Description & Specifications

Specifications

Chloroacetonitrile
100mL
107-14-2
Glass bottle
Liquid
98+%
ClCH2CN
MFCD00001885
Clear colorless
98+%
Authentic
1.4200 to 1.4240 (20°C, 589nm)
≥98.0%
C2H2ClN
75.5

Chemical Identifiers

107-14-2
C2H2ClN
75.5
Chloroacetonitrile, 98+%
98+%
ClCH2CN
MFCD00001885
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-467-0

RTECSNumber : AL8225000

TSCA : TSCA