Please login to your online account to display your discounted pricing

Chloroacetonitrile, 98+%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC108525000

 View more versions of this product

Catalog No. AC108525000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.0%
Formula Weight 75.5
Quantity 500mL
Color Clear colorless
TSCA TSCA
RTECS Number AL8225000
EINECS Number 203-467-0
Refractive Index 1.4200 to 1.4240 (20°C, 589nm)
Chemical Name or Material Chloroacetonitrile
Packaging Glass bottle
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C2H2ClN
Linear Formula ClCH2CN
MDL Number MFCD00001885
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 107-14-2
Health Hazard 1 Danger

Specifications

Chemical Name or Material Chloroacetonitrile
Name Note 98+%
CAS 107-14-2
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C2H2ClN
Linear Formula ClCH2CN
Formula Weight 75.5
MDL Number MFCD00001885
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

SDS & Safety Documents