Please login to your online account to display your discounted pricing

chloroacetic anhydride, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404421000

 View more versions of this product

Catalog No. AC404421000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color Brownish semi-transparent
Physical Form Crystals, Chunks or Low Melting Mass
Formula Weight 170.98
Quantity 100g
Beilstein 02, 199
Fieser 01,129
Merck Index 15, 2113
Melting Point 54° to 58°C
Chemical Name or Material chloroacetic anhydride
Linear Formula (ClCH2CO)2O
Assay Percent Range 97%
Molecular Formula C4H4Cl2O3
MDL Number MFCD00000929
CAS 541-88-8
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Contact with water liberates toxic gas.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material chloroacetic anhydride
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H4Cl2O3
Linear Formula (ClCH2CO)2O
Formula Weight 170.98
MDL Number MFCD00000929
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Contact with water liberates toxic gas.

SDS & Safety Documents