Please login to your online account to display your discounted pricing

Chloroacetic Acid Sodium Salt 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404415000

 View more versions of this product

Catalog No. AC404415000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 116.48
Quantity 500g
Color White
Merck Index 15, 2112
TSCA TSCA
EINECS Number 223-498-3
RTECS Number AG1400000
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Very toxic to aquatic life.
Name Note 98%
CAS 3926-62-3
Chemical Name or Material Chloroacetic acid sodium salt
Molecular Formula C2H2ClNaO2
MDL Number MFCD00002684
Assay Percent Range 98%
Linear Formula CH2ClCOONa
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Chloroacetic acid sodium salt
Name Note 98%
CAS 3926-62-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H2ClNaO2
Linear Formula CH2ClCOONa
Formula Weight 116.48
MDL Number MFCD00002684
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Very toxic to aquatic life.

SDS & Safety Documents