Please login to your online account to display your discounted pricing

Chloroacetic acid, ≥99%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC220320010

 View more versions of this product

Catalog No. AC220320010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99% (Acid)
Physical Form Liquid
Formula Weight 94.5
Quantity 1kg
Color Clear colorless
TSCA TSCA
EINECS Number 201-178-4
RTECS Number AF8575000
Water 0.3% Max.
Density 1.5800g/mL
Merck Index 15, 2112
Beilstein 02, 194
Infrared Spectrum Authentic
CAS 79-11-8
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00002683
Name Note p.a.
Linear Formula ClCH2CO2H
Assay Percent Range 99+%
Molecular Formula C2H3ClO2
Chemical Name or Material Chloroacetic acid
Packaging Glass bottle
Synonym MCA
Iron (Fe) 5ppm max.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chloroacetic acid
Name Note p.a.
CAS 79-11-8
Assay ≥99% (Acid)
Assay Percent Range 99+%
Molecular Formula C2H3ClO2
Linear Formula ClCH2CO2H
Formula Weight 94.5
MDL Number MFCD00002683
Synonym MCA
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

SDS & Safety Documents