missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroacetamidine hydrochloride, 96%

Manufacturer:  Alfa Aesar™ A1483503

 View more versions of this product

Catalog No. AAA1483503



Description

Description

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Chloroacetamidine hydrochloride
1g
C2H6Cl2N2
UN3261
chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
C(C(=N)N)Cl.Cl
128.984
129
97°C to 103°C
10300-69-3
MFCD00053013
3678354
GUPOZVHRTJYZCX-UHFFFAOYSA-N
2-chloroethanimidamide;hydrochloride
2776957
96%

Chemical Identifiers

10300-69-3
128.984
GUPOZVHRTJYZCX-UHFFFAOYSA-N
2776957
C(C(=N)N)Cl.Cl
C2H6Cl2N2
MFCD00053013
chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
2-chloroethanimidamide;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

SDS Documents