Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroacetaldehyde sodium bisulfite hydrate, 98% (dry wt.), water ca 5-6%

Chemical Identifiers

Chemical Name or Material Chloroacetaldehyde sodium bisulfite hydrate
CAS 13064-50-1
Molecular Formula C2H4ClNaO4S·xH2O
Formula Weight 182.56g/mol (Anhydrous)
MDL Number MFCD00059901
UN Number UN2811
Synonym 2-Chloro-1-hydroxyethanesulfonic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1270109

 
alfa aesar™
L12701-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Chemical Name or Material Chloroacetaldehyde sodium bisulfite hydrate
CAS 13064-50-1
Assay 98%
Assay Percent Notes (dry wt.), water ≈5 to 6%
Molecular Formula C2H4ClNaO4S·xH2O
Formula Weight 182.56g/mol (Anhydrous)
MDL Number MFCD00059901
UN Number UN2811
Synonym 2-Chloro-1-hydroxyethanesulfonic acid sodium salt
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.