Learn More
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2504936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chloroacetaldehyde dimethyl acetal | |
<-73°C | |
128°C to 130°C | |
Aromatic | |
1.415 | |
500 g | |
1733700 | |
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
COC(CCl)OC | |
124.56 | |
124.57 |
97-97-2 | |
1.094 | |
28°C (82°F) | |
C4H9ClO2 | |
MFCD00000948 | |
UN1993 | |
Moisture sensitive | |
CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
2-chloro-1,1-dimethoxyethane | |
60986 | |
98% |
Chemical Identifiers
97-97-2 | |
124.56 | |
CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
60986 | |
COC(CCl)OC |
C4H9ClO2 | |
MFCD00000948 | |
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
2-chloro-1,1-dimethoxyethane |
Safety and Handling
GHS H Statement
H301-H226-H332
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 202-624-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only