Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroacetaldehyde dimethyl acetal, 98%

Chemical Identifiers

Chemical Name or Material Chloroacetaldehyde dimethyl acetal
CAS 97-97-2
Formula Weight 124.57g/mol
MDL Number MFCD00000948
UN Number UN1993
Beilstein 1733700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3661614

 
alfa aesar™
36616-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3661622

 
alfa aesar™
36616-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Refractive Index 1.4150
Density 1.094g/mL
Melting Point <−73°C
Boiling Point 128° to 130°C
Quantity 25g
Flash Point 28° (82°F)
Chemical Name or Material Chloroacetaldehyde dimethyl acetal
CAS 97-97-2
Physical Form Liquid
Assay 98%
Formula Weight 124.57g/mol
MDL Number MFCD00000948
UN Number UN1993
Beilstein 1733700
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.