missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108502500

 View more versions of this product

Catalog No. AC108502500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloroacetaldehyde dimethyl acetal
97-97-2
Colorless to Yellow
128.0°C to 130.0°C
Authentic
Glass bottle
1.4140 to 1.4160
MFCD00000948
1.09
Solubility in water: 52g/L (20 c).
COC(CCl)OC
124.56
124.57
Liquid
97%
-73.0°C
1.0900g/mL
28°C
97%
C4H9ClO2
ClCH2CH(OCH3)2
250 mL
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
CRZJPEIBPQWDGJ-UHFFFAOYSA-N
2-chloro-1,1-dimethoxyethane
60986
97%

Chemical Identifiers

97-97-2
124.56
CRZJPEIBPQWDGJ-UHFFFAOYSA-N
60986
COC(CCl)OC
C4H9ClO2
MFCD00000948
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
2-chloro-1,1-dimethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-624-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only