Please login to your online account to display your discounted pricing

Chloroacetaldehyde Diethyl Acetal 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  108495000

 View more versions of this product

Catalog No. AC108495000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 152.62
Quantity 500mL
Color Clear colorless to slightly yellow
Fieser 05,107
Beilstein 01, 611
EINECS Number 210-695-4
TSCA TSCA
Refractive Index 1.4150 to 1.4170 (20°C, 589nm)
Assay Percent Range 99%
Chemical Name or Material Chloroacetaldehyde diethyl acetal
Molecular Formula C6H13ClO2
MDL Number MFCD00000949
Name Note 99%
CAS 621-62-5
Linear Formula ClCH2CH(OC2H5)2
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 2-Chloro-1, 1-diethoxyethane
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

Specifications

Chemical Name or Material Chloroacetaldehyde diethyl acetal
Name Note 99%
CAS 621-62-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H13ClO2
Linear Formula ClCH2CH(OC2H5)2
Formula Weight 152.62
MDL Number MFCD00000949
Synonym 2-Chloro-1, 1-diethoxyethane
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents