missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetaldehyde diethyl acetal, 99%, ACROS Organics™

$33.55 - $33.55

Chemical Identifiers

CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym 1,1-diethoxy-2-chloroethane, 2-chloroacetal, 2-chloroacetaldehyde diethyl acetal, acetaldehyde, chloro-, diethyl acetal, chloroacetal, chloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, diethoxyethyl chloride, ethane, 2-chloro-1,1-diethoxy, monochloroacetaldehyde diethyl acetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108490050
SDS View Documents
Thermo Fisher Scientific
108490050
5mL Glass bottle
Each for $33.55
 
Description

Description

Specifications

Chemical Identifiers

621-62-5
152.62
OVXJWSYBABKZMD-UHFFFAOYSA-N
12128
CCOC(CCl)OCC
C6H13ClO2
MFCD00000949
1,1-diethoxy-2-chloroethane, 2-chloroacetal, 2-chloroacetaldehyde diethyl acetal, acetaldehyde, chloro-, diethyl acetal, chloroacetal, chloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, diethoxyethyl chloride, ethane, 2-chloro-1,1-diethoxy, monochloroacetaldehyde diethyl acetal
2-chloro-1,1-diethoxyethane

Specifications

1.0100g/mL
Authentic
1.4150 to 1.4170
157.0°C
-45.0°C
621-62-5
C6H13ClO2
MFCD00000949
05,107
Solubility in water: 1.8g/L (20 c).
CCOC(CCl)OCC
152.62
152.62
99%
29°C
Glass bottle
1.01
Colorless to Yellow
5mL
99%
ClCH2CH(OC2H5)2
01, 611
1,1-diethoxy-2-chloroethane, 2-chloroacetal, 2-chloroacetaldehyde diethyl acetal, acetaldehyde, chloro-, diethyl acetal, chloroacetal, chloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, diethoxyethyl chloride, ethane, 2-chloro-1,1-diethoxy, monochloroacetaldehyde diethyl acetal
OVXJWSYBABKZMD-UHFFFAOYSA-N
2-chloro-1,1-diethoxyethane
12128
Liquid
Chloroacetaldehyde diethyl acetal, 99%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-695-4

TSCA : TSCA

SDS
Product Certifications