missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetaldehyde diethyl acetal, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 108490050

 View more versions of this product

Catalog No. AC108490050



Description

Description

Specifications

Specifications

Chloroacetaldehyde diethyl acetal
621-62-5
Colorless to Yellow
157.0°C
Authentic
Glass bottle
1.4150 to 1.4170
MFCD00000949
01, 611
1.01
Solubility in water: 1.8g/L (20 c).
CCOC(CCl)OCC
152.62
152.62
Liquid
99%
-45.0°C
1.0100g/mL
29°C
99%
C6H13ClO2
ClCH2CH(OC2H5)2
5 mL
05,107
chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
OVXJWSYBABKZMD-UHFFFAOYSA-N
2-chloro-1,1-diethoxyethane
12128
99%

Chemical Identifiers

621-62-5
152.62
OVXJWSYBABKZMD-UHFFFAOYSA-N
12128
CCOC(CCl)OCC
C6H13ClO2
MFCD00000949
chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
2-chloro-1,1-diethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-695-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only