Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroacetaldehyde diethyl acetal, 98%

Chemical Identifiers

Chemical Name or Material Chloroacetaldehyde diethyl acetal
CAS 621-62-5
Molecular Formula C6H13ClO2
Linear Formula ClCH2CH(OCH2CH3)2
Formula Weight 152.62
MDL Number MFCD00000949
UN Number UN1993
Beilstein 969254
Synonym 2-Chloro-1,1-diethoxyethane; Diethyl chloroacetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0766122

  SDS
alfa aesar™
L0766122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0766136

  SDS
alfa aesar™
L0766136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.62
CAS 621-62-5
MDL Number MFCD00000949
Chemical Name or Material Chloroacetaldehyde diethyl acetal
Molecular Formula C6H13ClO2
Synonym 2-Chloro-1,1-diethoxyethane; Diethyl chloroacetal
Beilstein 969254
Assay 98%
Health Hazard 1 Danger
UN Number UN1993
TSCA TSCA
Quantity 100g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Melting Point −45°C
Boiling Point 156° to 157°C
Flash Point 50°C (122°F)
Linear Formula ClCH2CH(OCH2CH3)2
Health Hazard 2 GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.