Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroacetaldehyde, 45% w/w in water

$33.76 - $33.76

Chemical Identifiers

Chemical Name or Material Chloroacetaldehyde
CAS 107-20-0
Molecular Formula C2H3ClO
Linear Formula ClCH2CHO
Formula Weight 78.5
MDL Number MFCD00006992
UN Number UN2232
Beilstein 1071226
Merck Index 14,2109
Synonym 2-Chloroethanal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1103730

  SDS
alfa aesar™
A1103730
250g Each for $33.76

AAA110370B

  SDS
alfa aesar™
A110370B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chloroacetaldehyde
Concentration 45% w/w in water
Flash Point 62°C (143°F)
Boiling Point 80° to 100°C
Melting Point −28°C
Quantity 250g
pH 1.2
CAS 107-20-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H330-H314-H318-H351-H227
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: 2-CHLOROETHANAL
RTECS Number AB2450000
TSCA TSCA
Odor Pungent
Molecular Formula C2H3ClO
Linear Formula ClCH2CHO
Formula Weight 78.5
MDL Number MFCD00006992
UN Number UN2232
Beilstein 1071226
Merck Index 14,2109
Synonym 2-Chloroethanal

Applications
Chloroacetaldehyde is a useful intermediate in the synthesis, it is also used to facilitate bark removal from tree trunks.

Solubility
soluble in acetone, methanol. Fully miscible in water.

Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.