Please login to your online account to display your discounted pricing

Chloro(triphenylphosphine)gold(I), 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391612500

 View more versions of this product

Catalog No. AC391612500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex Au)
Physical Form Crystalline Powder
Formula Weight 494.71
Quantity 250mg
Color White
Chemical Name or Material Chloro(triphenylphosphine)gold(I)
Packaging Glass bottle
CAS 14243-64-2
Assay Percent Range 99%
Molecular Formula C18H15AuClP
MDL Number MFCD00009588
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Chloro(triphenylphosphine)gold(I)
CAS 14243-64-2
Assay ≥98.5% (ex Au)
Assay Percent Range 99%
Molecular Formula C18H15AuClP
Formula Weight 494.71
MDL Number MFCD00009588
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents