Please login to your online account to display your discounted pricing

α'-Chloro-α,α,α-trifluoro-m-xylene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α'-Chloro-α, α, α-trifluoro-m-xylene
CAS 705-29-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6ClF3
Linear Formula ClCH2C6H4CF3
Formula Weight 194.58
MDL Number MFCD00000908
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC173510250

  SDS
thermo fisher scientific
173510250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 194.58
Quantity 25g
Color Clear colorless to slightly yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material α'-Chloro-α, α, α-trifluoro-m-xylene
Assay Percent Range 97%
CAS 705-29-3
Molecular Formula C8H6ClF3
Linear Formula ClCH2C6H4CF3
MDL Number MFCD00000908
Refractive Index 1.4630 to 1.4650 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.