Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)

Chemical Identifiers

Chemical Name or Material Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
CAS 92361-49-4
Formula Weight 796.33g/mol
MDL Number MFCD09038426
Synonym Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp&asterisk;(PPh{3})2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4662903

 
alfa aesar™
46629-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Melting Point 128°C (dec.)
Color Yellow
Quantity 1g
Sensitivity Air Sensitive
Chemical Name or Material Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
CAS 92361-49-4
Physical Form Crystalline Solid
Formula Weight 796.33g/mol
MDL Number MFCD09038426
Synonym Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride; RuClCp&asterisk;(PPh{3})2
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C