Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I), 97%

$177.80 - $644.89

Chemical Identifiers

Chemical Name or Material Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I)
CAS 213603-12-4
Molecular Formula C23H33ClN2O2RuS
Formula Weight 538.11
MDL Number MFCD12546183
Synonym [(R,R)-TsDACH RuCl(p-cymene)]; (p-Cymene)N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3026403

  SDS
alfa aesar™
H3026403
1g Each for $644.89

AAH30264MD

  SDS
alfa aesar™
H30264MD
250mg Each for $177.80
Description & Specifications

Specifications

Formula Weight 538.11
Molecular Formula C23H33ClN2O2RuS
Quantity 1g
Synonym [(R,R)-TsDACH RuCl(p-cymene)]; (p-Cymene)N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I) chloride
Chemical Name or Material Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I)
MDL Number MFCD12546183
Assay 97%
CAS 213603-12-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.