missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloro(dimethyl)thexylsilane, TCI America™

Manufacturer:  TCI America T21165G

 View more versions of this product

Catalog No. T21165G



Specifications

Specifications

Chloro(dimethyl)thexylsilane
Colorless
67373-56-2
MFCD00012175
dimethylthexylsilyl chloride, thexyldimethylchlorosilane, chloro dimethyl thexylsilane, chloro 2,3-dimethylbutan-2-yl dimethylsilane, dimethylthexylchlorosilane, thexyldimethylsilyl chloride, chlorodimethyl 1,1,2-trimethylpropyl silane, dimethylthexylsilylchloride, tds-cl, acmc-209nyu
CC(C)C(C)(C)[Si](C)(C)Cl
178.775
178.78
≥95.0% (GC)
56°C
5g
C8H19ClSi
2986
KIGALSBMRYYLFJ-UHFFFAOYSA-N
chloro-(2,3-dimethylbutan-2-yl)-dimethylsilane
144239
Clear Liquid at 20°C

Chemical Identifiers

67373-56-2
178.775
KIGALSBMRYYLFJ-UHFFFAOYSA-N
144239
CC(C)C(C)(C)[Si](C)(C)Cl
C8H19ClSi
MFCD00012175
dimethylthexylsilyl chloride, thexyldimethylchlorosilane, chloro dimethyl thexylsilane, chloro 2,3-dimethylbutan-2-yl dimethylsilane, dimethylthexylchlorosilane, thexyldimethylsilyl chloride, chlorodimethyl 1,1,2-trimethylpropyl silane, dimethylthexylsilylchloride, tds-cl, acmc-209nyu
chloro-(2,3-dimethylbutan-2-yl)-dimethylsilane
Safety and Handling

Safety and Handling

TSCA : No

Documents