Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(dimethyl)phosphine, 97%

Chemical Identifiers

Chemical Name or Material Chloro(dimethyl)phosphine
CAS 811-62-1
Linear Formula (CH3)2PCl
Formula Weight 96.5
MDL Number MFCD00013620
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3016002

  SDS
alfa aesar™
3016002
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.5
Linear Formula (CH3)2PCl
Health Hazard 2 GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 76° to 77°C
Flash Point −18° (−1°F)
Sensitivity Air & Moisture Sensitive
Density 1.22g/mL
Odor Pungent
Assay 97%
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
UN Number UN2845
MDL Number MFCD00013620
Health Hazard 1 Danger
CAS 811-62-1
Color Colorless
Quantity 0.5g
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Chloro(dimethyl)phosphine
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.