missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

chloro(diethyl)phosphine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC350310050

 View more versions of this product

Catalog No. AC350310050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Chloro(diethyl)phosphine
Clear colorless to light yellow
686-69-1
94% min.
1.4740 to 1.4760 (20°C, 589nm)
95%
(CH3CH2)2PCl
MFCD00013621
1.0230g/mL
5g
Authentic
Glass bottle
Liquid
C4H10ClP
124.55

Chemical Identifiers

95%
(CH3CH2)2PCl
MFCD00013621
C4H10ClP
124.55
Chloro(diethyl)phosphine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger