Please login to your online account to display your discounted pricing

chloro(diethyl)phosphine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chloro(diethyl)phosphine
Assay Percent Range 95%
Molecular Formula C4H10ClP
Linear Formula (CH3CH2)2PCl
Formula Weight 124.55
MDL Number MFCD00013621
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350310050

  SDS
thermo fisher scientific
AC350310050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350310010

  SDS
thermo fisher scientific
350310010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 124.55
Quantity 5g
Color Clear colorless to light yellow
Density 1.0230g/mL
Chemical Name or Material Chloro(diethyl)phosphine
Molecular Formula C4H10ClP
Linear Formula (CH3CH2)2PCl
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
CAS 686-69-1
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
NMR 94% min.
MDL Number MFCD00013621
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.