Please login to your online account to display your discounted pricing

chloro(diethyl)phosphine, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350310010

 View more versions of this product

Catalog No. AC350310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 124.55
Quantity 1g
Color Clear colorless to light yellow
Linear Formula (CH3CH2)2PCl
Infrared Spectrum Authentic
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
MDL Number MFCD00013621
Assay Percent Range 95%
NMR 94% min.
Molecular Formula C4H10ClP
Packaging Glass bottle
Chemical Name or Material Chloro(diethyl)phosphine
CAS 686-69-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material Chloro(diethyl)phosphine
Assay Percent Range 95%
Molecular Formula C4H10ClP
Linear Formula (CH3CH2)2PCl
Formula Weight 124.55
MDL Number MFCD00013621
Accessories

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents