Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(diethyl)phosphine

Chemical Identifiers

Chemical Name or Material Chloro(diethyl)phosphine
CAS 686-69-1
Linear Formula (C2H5)2PCl
Formula Weight 124.55
MDL Number MFCD00013621
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3020403

  SDS
alfa aesar™
3020403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3020406

  SDS
alfa aesar™
3020406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.55
Chemical Name or Material Chloro(diethyl)phosphine
Refractive Index 1.4750
Physical Form Liquid
Linear Formula (C2H5)2PCl
MDL Number MFCD00013621
CAS 686-69-1
UN Number UN2845
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Density 1.023g/mL
Flash Point 18° (66°F)
Sensitivity Air & Moisture Sensitive
Boiling Point 131° to 132°C
Color Colorless to pale yellow
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.