missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(diethyl)phosphine

Manufacturer:  Alfa Aesar™ 3020403

 View more versions of this product

Catalog No. AA3020403



Specifications

Specifications

Chloro(diethyl)phosphine
18° (66°F)
Air & Moisture Sensitive
Colorless to Yellow
686-69-1
(C2H5)2PCl
UN2845
INJBDKCHQWVDGT-UHFFFAOYSA-N
chloro(diethyl)phosphane
69639
Liquid
1.023g/mL
1.4750
131°C to 132°C
1g
C4H10ClP
MFCD00013621
chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
CCP(CC)Cl
124.548
124.55

Chemical Identifiers

686-69-1
124.548
INJBDKCHQWVDGT-UHFFFAOYSA-N
69639
CCP(CC)Cl
C4H10ClP
MFCD00013621
chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
chloro(diethyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

SDS Documents