Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)

Chemical Identifiers

Chemical Name or Material Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
CAS 1252598-33-6
Formula Weight 907.69
MDL Number MFCD25372545
Synonym PdCl(crotyl)QPhos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4663903

 
alfa aesar™
4663903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 907.69
Chemical Name or Material Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Physical Form Solid
MDL Number MFCD25372545
CAS 1252598-33-6
Synonym PdCl(crotyl)QPhos
Color Pink
Quantity 1g