Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloro(2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl)(2'-aminobiphenyl-2-yl)palladium(II)

Chemical Identifiers

Chemical Name or Material Chloro(2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl)(2'-aminobiphenyl-2-yl)palladium(II)
CAS 1310584-14-5
Formula Weight 786.80g/mol
MDL Number MFCD20264878
Synonym XPhos Palladacycle; 2nd Generation XPhos Precatalyst
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4717403

 
alfa aesar™
47174-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color White
Quantity 1g
Chemical Name or Material Chloro(2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl)(2'-aminobiphenyl-2-yl)palladium(II)
CAS 1310584-14-5
Physical Form Powder
Formula Weight 786.80g/mol
MDL Number MFCD20264878
Synonym XPhos Palladacycle; 2nd Generation XPhos Precatalyst
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

  • Buchwald-Hartwig aminations and Suzuki-Miyaura cross-coupling reactions

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.