missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC211300050

 View more versions of this product

Catalog No. AC211300050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Chloro(1, 5-cyclooctadiene)rhodium(I) dimer
Glass bottle
256.0°C
min. 40.8% Rh
C16H24Cl2Rh2
1,5-cyclooctadiene rhodium chloride dimer, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, ccris 5036, chloro 1,5-cyclooctadiene rhodium i dimer, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride
PDJQCHVMABBNQW-MIXQCLKLSA-L
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride
6436379
Crystals or Crystalline Powder
Authentic
Undesignated
5g
12092-47-6
MFCD00012415
Solubility in water: 0.34g/L
C1CC=CCCC=C1.C1CC=CCCC=C1.[Cl-].[Cl-].[Rh].[Rh]
493.08
493.08
≥40.8% (RH)

Chemical Identifiers

12092-47-6
493.08
PDJQCHVMABBNQW-MIXQCLKLSA-L
6436379
C1CC=CCCC=C1.C1CC=CCCC=C1.[Cl-].[Cl-].[Rh].[Rh]
C16H24Cl2Rh2
MFCD00012415
1,5-cyclooctadiene rhodium chloride dimer, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, ccris 5036, chloro 1,5-cyclooctadiene rhodium i dimer, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 235-157-6

TSCA : TSCA

SDS Documents