missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), Alfa Aesar™

Manufacturer:  Alfa Aesar 4680303

 View more versions of this product

Catalog No. AA4680303



Specifications

Specifications

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
143°C to 147°C
92390-26-6
UN3395
379.94
Brown-Yellow
1g
MFCD07369035
Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride; RuCl(cod)Cp*
Crystalline Solid

Chemical Identifiers

92390-26-6
MFCD07369035
Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride; RuCl(cod)Cp*
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE

storageNote1 : -30° to -10°C

SDS Documents