Please login to your online account to display your discounted pricing

Chloro(1,5-cyclooctadiene)iridium(I) dimer, White crystalline powder, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Chloro(1, 5-cyclooctadiene)iridium(I) dimer
CAS 12112-67-3
Physical Form Crystalline Powder
Molecular Formula C16H24Cl2Ir2
Formula Weight 671.71
MDL Number MFCD00012414
Synonym Bis(1, 5-cyclooctadiene)diiridium(I) dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1274901

  SDS
alfa aesar
1274901
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1274903

  SDS
alfa aesar
1274903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1274906

  SDS
alfa aesar
1274906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 671.71
Physical Form Crystalline Powder
Color White
Quantity 0.25g
Molecular Formula C16H24Cl2Ir2
Odor Odorless
Synonym Bis(1, 5-cyclooctadiene)diiridium(I) dichloride
MDL Number MFCD00012414
Chemical Name or Material Chloro(1, 5-cyclooctadiene)iridium(I) dimer
CAS 12112-67-3