Please login to your online account to display your discounted pricing

Chlormethiazole hydrochloride, Tocris


Potentiates GABAA receptor function

Quantity 50mg

Manufacturer: r & d systems  RDS088150

 View more versions of this product

Catalog No. 088150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 198.11
Molecular Formula C6H8ClNS·HCl
CAS 6001-74-7
Assay Percent Range >99%
Chemical Name or Material Chlormethiazole hydrochloride
Synonym 5-(2-Chloroethyl)-4-methylthiazole hydrochloride
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material Chlormethiazole hydrochloride
Assay Percent Range >99%
Molecular Formula C6H8ClNS·HCl
Formula Weight 198.11
Synonym 5-(2-Chloroethyl)-4-methylthiazole hydrochloride
Accessories

Desiccate at Room Temperature