Please login to your online account to display your discounted pricing

Chlormethiazole hydrochloride, Tocris

Potentiates GABAA receptor function

$155.12 - $641.36

Chemical Identifiers

Chemical Name or Material Chlormethiazole hydrochloride
Assay Percent Range >99%
Molecular Formula C6H8ClNS·HCl
Formula Weight 198.11
Synonym 5-(2-Chloroethyl)-4-methylthiazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

088110

 
r & d systems
RDS088110
10mg Each for $155.12

088150

 
r & d systems
RDS088150
50mg Each for $641.36
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 198.11
Chemical Name or Material Chlormethiazole hydrochloride
Assay Percent Range >99%
Molecular Formula C6H8ClNS·HCl
CAS 6001-74-7
Synonym 5-(2-Chloroethyl)-4-methylthiazole hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature