missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlormethiazole Hydrochloride, TCI America™

Manufacturer:  TCI America C30201G

 View more versions of this product

Catalog No. C30201G



Specifications

Specifications

Chlormethiazole Hydrochloride
136°C
6001-74-7
MFCD00673948
OFXYKSLKNMTBHK-UHFFFAOYSA-N
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
19035073
Crystal-Powder at 20°C
White
1g
C6H9Cl2NS
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
CC1=C(SC=N1)CCCl.Cl
198.105
198.11
≥98.0% (HPLC,T)

Chemical Identifiers

6001-74-7
198.105
OFXYKSLKNMTBHK-UHFFFAOYSA-N
19035073
CC1=C(SC=N1)CCCl.Cl
C6H9Cl2NS
MFCD00673948
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents