Please login to your online account to display your discounted pricing

Chlorisondamine diiodide, Tocris


Nicotinic antagonist; slow offset

Quantity 50mg

Manufacturer: r & d systems  100150

 View more versions of this product

Catalog No. 100150

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 611.95
Quantity 50mg
Assay Percent Range >99%
Chemical Name or Material Chlorisondamine diiodide
Molecular Formula C14H20Cl4I2N2
CAS 96750-66-2
Synonym 4,5,6,7-Tetrachloro-1,3-dihydro-2-methyl-2-[2-trimethylammonium)ethyl]-2H-isoindolium diiodide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Chlorisondamine diiodide
Assay Percent Range >99%
Molecular Formula C14H20Cl4I2N2
Formula Weight 611.95
Synonym 4,5,6,7-Tetrachloro-1,3-dihydro-2-methyl-2-[2-trimethylammonium)ethyl]-2H-isoindolium diiodide
Accessories

Desiccate at +4°C