Please login to your online account to display your discounted pricing

Chlorisondamine diiodide, Tocris

Nicotinic antagonist; slow offset

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Chlorisondamine diiodide
Assay Percent Range >99%
Molecular Formula C14H20Cl4I2N2
Formula Weight 611.95
Synonym 4,5,6,7-Tetrachloro-1,3-dihydro-2-methyl-2-[2-trimethylammonium)ethyl]-2H-isoindolium diiodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

100110

 
r & d systems
100110
10mg Each for $177.37

100150

 
r & d systems
100150
50mg Each for $746.48
Description & Specifications

Specifications

Chemical Name or Material Chlorisondamine diiodide
Quantity 10mg
CAS 96750-66-2
Recommended Storage Desiccate at +4°C
Assay Percent Range >99%
Molecular Formula C14H20Cl4I2N2
Formula Weight 611.95
Synonym 4,5,6,7-Tetrachloro-1,3-dihydro-2-methyl-2-[2-trimethylammonium)ethyl]-2H-isoindolium diiodide
Safety and Handling

Desiccate at +4°C