Please login to your online account to display your discounted pricing

Chlorinated Herbicides Mixture, 6 Analytes, Various concentrations in Methanol, Ultra Scientific


Quantity 1 x 1mL

Manufacturer: ultra scientific  HBM5152M1

 View more versions of this product

Catalog No. US-HBM-5152M1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1 x 1mL
Physical Form Liquid
Chemical Name or Material Chlorinated Herbicides Mixture
CAS 000120-36-5
CAS 000087-86-5
CAS 000067-56-1
Concentration or Composition (by Analyte or Components) Tetrachloroterephthalic acid (dacthal acid metabolite): 100μg/mL, 3,5-dichlorobenzoic acid: 500μg/mL, dichlorprop: 100μg/mL, dinoseb: 200μg/mL, pentachlorophenol: 100μ
CAS 002136-79-0
CAS 000088-85-7
CAS 000051-36-5

Specifications

Chemical Name or Material Chlorinated Herbicides Mixture
CAS 000067-56-1
CAS 002136-79-0
CAS 000051-36-5
CAS 000120-36-5
CAS 000088-85-7
CAS 000087-86-5
Physical Form Liquid