Chlorinated Herbicides Mixture, 6 Analytes, Various concentrations in Methanol, Ultra Scientific
Manufacturer:  Ultra Scientific HBM5152A1
 View more versions of this product
Catalog No. US-HBM-5152A1
Specifications Description & Specifications
Specifications
1 x 1mL
Tetrachloroterephthalic acid (dacthal acid metabolite): 100μg/mL, 3,5-dichlorobenzoic acid: 500μg/mL, dichlorprop: 100μg/mL, dinoseb: 200μg/mL, pentachlorophenol: 100μ
Chlorinated Herbicides Mixture
Liquid
000087-86-5,000120-36-5,002136-79-0,000051-36-5,000067-56-1,000088-85-7
Chemical Identifiers
000067-56-1,002136-79-0,000051-36-5,000120-36-5,000088-85-7,000087-86-5
Liquid
Chlorinated Herbicides Mixture