Please login to your online account to display your discounted pricing

Chlorinated Herbicides Mixture, 6 Analytes, Various concentrations in Methanol, Ultra Scientific

$221.14 - $410.60

Chemical Identifiers

Chemical Name or Material Chlorinated Herbicides Mixture
CAS 000067-56-1
CAS 002136-79-0
CAS 000051-36-5
CAS 000120-36-5
CAS 000088-85-7
CAS 000087-86-5
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-HBM-5152A1

 
ultra scientific
HBM5152A1
1 x 1mL Each for $221.14

US-HBM-5152M1

 
ultra scientific
HBM5152M1
1 x 1mL Each for $410.60
Description & Specifications

Specifications

Quantity 1 x 1mL
Physical Form Liquid
Chemical Name or Material Chlorinated Herbicides Mixture
CAS 000120-36-5
CAS 000087-86-5
Concentration or Composition (by Analyte or Components) Tetrachloroterephthalic acid (dacthal acid metabolite): 100μg/mL, 3,5-dichlorobenzoic acid: 500μg/mL, dichlorprop: 100μg/mL, dinoseb: 200μg/mL, pentachlorophenol: 100μ
CAS 002136-79-0
CAS 000051-36-5
CAS 000088-85-7
CAS 000067-56-1