missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloride Standard, 1mL=0.05mg Cl, 50ppm Cl, Ricca Chemical

$43.12 - $74.45

Chemical Identifiers

CAS 7732-18-5
Physical Form Liquid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
194516 SDS Ricca Chemical Company
RI194516
500mL Each for $74.45
R1945000120 Ricca Chemical Company
RIR1945000120A
Poly natural 120mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R19450001A Ricca Chemical Company
RIR19450001A
Poly natural 1L Each for $70.55
R19450004A Ricca Chemical Company
RIR19450004A
Poly natural 4L Each for $43.12
Specifications Description & Specifications

Chemical Identifiers

7732-18-5
Liquid

Specifications

500mL
Liquid
Clear colorless liquid
Odorless
Solubility in Water: Infinite
7732-18-5
6.7 to 7.3

Safety and Handling

Recommended Storage : General