missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloride Standard, 1mL=0.05mg Cl, 50ppm Cl, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR19450001A

 View more versions of this product

Catalog No. R19450001A



Specifications

Specifications

7732-18-5
Liquid
6.7 to 7.3
Solubility in Water: Infinite
Clear colorless liquid
Odorless
Poly natural
1L

Chemical Identifiers

7732-18-5
Safety and Handling

Safety and Handling

Recommended Storage : General

Documents