missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloride Standard, 1mL = 0.005mg Cl-, 5ppm Cl-, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR1941500120A

 View more versions of this product

Catalog No. R1941500120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Chloride Standard
7647-14-5,7732-18-5
ClNa
[Na+].[Cl-]
58.44
CHEBI:26710
Odorless
Solubility in Water: Infinite
Colorless
1mL = 0.005mg Cl-, 5ppm Cl-
5.00 ± 0.05ppm
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Liquid
Poly natural
Aqueous Solution
120mL

Chemical Identifiers

7647-14-5
58.44
5234
sodium;chloride
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
Documents