Please login to your online account to display your discounted pricing

Chloride Standard, 1mL = 0.001mg Cl-, 1ppm Cl-, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Chloride Standard
Name Note 1mL = 0.001mg Cl-, 1ppm Cl-
CAS 7647-14-5
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 1.00 ± 0.01ppm
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R1941100120

 
ricca chemical company
RIR1941100120A
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R19411001A

 
ricca chemical company
RIR19411001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R19411004A

 
ricca chemical company
RIR19411004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 120mL
Chemical Name or Material Chloride Standard
CAS 7647-14-5
Assay Percent Range 1.00 ± 0.01ppm
Name Note 1mL = 0.001mg Cl-, 1ppm Cl-
Solvent or Matrix Aqueous Solution
Solubility Solubility in Water: Infinite
Odor Odorless
Packaging Poly natural
CAS 7732-18-5