missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloride Standard, 1000ppm, Ricca Chemical

$53.33 - $156.48

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1955-4
SDS View Documents
Ricca Chemical Company
RI19554
120mL Poly natural
Each for $54.42
 
195516
SDS View Documents
Ricca Chemical Company
RI195516*
500mL Poly bottle
Each for $120.33
 
SC023100500
View Documents
Ricca Chemical Company
RISC023100500A
500mL Poly natural
Each for $156.48
 
R19550001A
View Documents
Ricca Chemical Company
RIR19550001A
1L Poly natural
Each for $80.12
 
R19550004A
View Documents
Ricca Chemical Company
RIR19550004A
4L Poly natural
Each for $53.33
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
5234
sodium;chloride
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]

Specifications

7647-14-5,7732-18-5
100.0,0.17
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Liquid
1000ppm
Colorless
Chloride Standard
98.0,0.16
ClNa
[Na+].[Cl-]
58.44
CHEBI:26710
Poly natural
1 mL = 1 mg Cl
120mL
SDS