missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloride Standard, 1000ppm, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR19550001A

 View more versions of this product

Catalog No. R19550001A



Specifications

Specifications

Chloride Standard
98.0,0.16
ClNa
[Na+].[Cl-]
58.44
CHEBI:26710
Poly natural
1 mL = 1 mg Cl
1L
7647-14-5,7732-18-5
100.0,0.17
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Liquid
1000ppm
Colorless

Chemical Identifiers

7647-14-5
58.44
5234
sodium;chloride
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
Documents